Learn More
Emma Safety Footwear Zandvoort Safety Shoes
Click to view available options
Size:
37
38
39
40
41
42
43
46
47
48
Unit Size:
1 pair
Description
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Duo Heat Resistant Outsole
- Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345:2011 |
| Color | Brown |
| Design | Zandvoort |
| Liner Material | Full grain Nappa leather / Split leather |
| Material | Oiled Nubuck |
| Size | 42 |