missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Z,Z)-5,11-Eicosadienoic Acid Methyl Ester-d3, TRC
Specifications
Specifications
| Molecular Formula | C23 H48 . C13 H28 O4 S4 . C11 H20 O3 . 7 C H4 |
| Quantity | 1 mg |
| Synonym | (5Z,11Z)-5,11-Eicosadienoic Acid Methyl Ester-d3, (5Z,11Z)-Eicosadienoic Acid Methyl Ester-d3, Keteleeronic Acid Methyl Ester-d3, all-cis-5,11-Eicosadienoic Acid Methyl Ester-d3 |
| InChI Formula | InChI=1S/C23H48.C13H28O4S4.C11H20O3.7CH4/c1-3-5-7-9-11-13-15-17-19-21-23-22-20-18-16-14-12-10-8-6-4-2;1-20(14,15)18-12-10-8-6-4-3-5-7-9-11-13-19-21(2,16)17;1-9-10(14-9)7-5-3-2-4-6-8-11(12)13;;;;;;;/h3-23H2,1-2H3;3-13H2,1-2H3;9-10H,2-8H2,1H3,(H,12,13);7*1H4 |
| SMILES | C.C.C.C.C.C.C.CCCCCCCCCCCCCCCCCCCCCCC.CC1OC1CCCCCCCC(=O)O.CS(=O)(=O)SCCCCCCCCCCCSS(=O)(=O)C |
| IUPAC Name | 1,11-bis(methylsulfonylsulfanyl)undecane;methane;8-(3-methyloxiran-2-yl)octanoic acid;tricosane |
| Molecular Weight (g/mol) | 1013.818 |
| Formula Weight | 1012.823 |
| Chemical Name or Material | (Z,Z)-5,11-Eicosadienoic Acid Methyl Ester-d3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?