Learn More
Z-Gly-Arg-thiobenzyl Ester, MP Biomedicals™
Product Code. p-7236188 Shop All MP Biomedical Products
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 11411025
Brand: MP Biomedicals 03SB00710
Z-Gly-Arg-thiobenzyl Ester is a chromogenic substrate for Plasmogen Activator Serine Protease.
  • Peptide thiobenzyl esters are sensitive substrates for serine and cysteine proteases.
  • The thiol leaving group is detected by reaction with a thiol reagent (DTNB or Leman's reagent) or 4,4'-dithiodipyridine.
  • The rate of thiobenzyl ester hydrolysis is measured at 324 nm by monitoring the formation of 4-thiopyridone (E324nm = 19,800 M-1cm-1) or at 405 nm using DTNB as a thiol reagent (E405nm = 13,260 M-1cm-1) for 2-nitro-5-thiobenzoate anion TNBS.
  • Protein or Enzyme Type: Activators, Inhibitors and Substrates
TRUSTED_SUSTAINABILITY
Specifications
Form Semi-solid
Quantity 10 mg
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.