missing translation for 'onlineSavingsMsg'
Learn More

Z-Gly-Arg-thiobenzyl Ester, MP Biomedicals™

Product Code. 11411025 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411025 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11411025 Supplier MP Biomedicals Supplier No. 03SB00710
  • ONLINE EXCLUSIVE PRICE
    772.00€
    735.00€ / 10mg
    Save 37.00€
    4% Off
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Z-Gly-Arg-thiobenzyl Ester is a chromogenic substrate for Plasmogen Activator Serine Protease.

  • Peptide thiobenzyl esters are sensitive substrates for serine and cysteine proteases.
  • The thiol leaving group is detected by reaction with a thiol reagent (DTNB or Leman's reagent) or 4,4'-dithiodipyridine.
  • The rate of thiobenzyl ester hydrolysis is measured at 324 nm by monitoring the formation of 4-thiopyridone (E324nm = 19,800 M-1cm-1) or at 405 nm using DTNB as a thiol reagent (E405nm = 13,260 M-1cm-1) for 2-nitro-5-thiobenzoate anion TNBS.
  • Protein or Enzyme Type: Activators, Inhibitors and Substrates
TRUSTED_SUSTAINABILITY

Specifications

Form Semi-solid
Quantity 10 mg

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.