missing translation for 'onlineSavingsMsg'
Learn More

(Z)-2-(5-Amino-1,2,4-thiadiazol-3-yl)-2-(((1-(tert-butoxy)-2-methyl-1-oxopropan-2-yl)oxy)imino)acetic Acid, TRC

Product Code. 30336201
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336201 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336201 Supplier Toronto Research Chemicals Supplier No. TRCA8713901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (Z)-2-(5-Amino-1,2,4-thiadiazol-3-yl)-2-(((1-(tert-butoxy)-2-methyl-1-oxopropan-2-yl)oxy)imino)acetic Acid
CAS 76028-96-1
Molecular Formula C12 H18 N4 O5 S
Quantity 1 g
Synonym 1,2,4-Thiadiazole-3-acetic acid, 5-amino-α-[[2-(1,1-dimethylethoxy)-1,1-dimethyl-2-oxoethoxy]imino]-, (Z)-, (αZ)-5-Amino-α-[[2-(1,1-dimethylethoxy)-1,1-dimethyl-2-oxoethoxy]imino]-1,2,4-thiadiazole-3-acetic acid, (Z)-2-(5-Amino-1,2,4-thiadiazol-3-yl)-2-(((1-(tertbutoxy)-2-methyl-1-oxopropan-2-yl)oxy)imino) acetic acid, (Z)-2-(5-Amino-1,2,4-thiadiazol-3-yl)-2-(1-(1,1-dimethylethoxy)carbonyl-1-methylethoxyimino)acetic acid
InChI Formula InChI=1S/C12H18N4O5S/c1-11(2,3)20-9(19)12(4,5)21-15-6(8(17)18)7-14-10(13)22-16-7/h1-5H3,(H,17,18)(H2,13,14,16)/b15-6-
SMILES CC(C)(C)OC(=O)C(C)(C)O\N=C(/C(=O)O)\c1nsc(N)n1
IUPAC Name (2Z)-2-(5-amino-1,2,4-thiadiazol-3-yl)-2-[2-methyl-1-[(2-methylpropan-2-yl)oxy]-1-oxopropan-2-yl]oxyiminoacetic acid
Molecular Weight (g/mol) 330.36
Formula Weight 330.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.