missing translation for 'onlineSavingsMsg'
Learn More

YttriumTrifluoroacetate (~90%), TRC

Product Code. 30454137
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454137 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454137 Supplier Toronto Research Chemicals Supplier No. TRCY1200401MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material YttriumTrifluoroacetate
CAS 37737-28-3
Molecular Formula C6H2F9O7Y
Quantity 1 mg
Synonym Tris(trifluoroacetato)yttrium, Yttrium tris(trifluoroacetate)
InChI Formula InChI=1S/C2HF3O2.Y/c3-2(4,5)1(6)7;/h(H,6,7);/q;+3/p-1
SMILES [Y+3].[O-]C(=O)C(F)(F)F
IUPAC Name 2,2,2-trifluoroacetate;yttrium(3+)
Molecular Weight (g/mol) 445.97
Formula Weight 201.8909
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.