Learn More
YSI™ Fill Conductivity Cell
Click to view available options
Connector Types:
7-pin Mini-DIN
Unit Size:
Each
Specifications
Specifications
| Cell Constant | 1 cm-1 |
| Connector Types | 7-pin Mini-DIN |
| For Use With (Equipment) | 3200 and 3400 Meter |
| Diameter (English) Body | 0.75 in. |
| Diameter (Metric) Body | 19.05 mm |
| Length (English) | 5.25 in. |
| Length (English) Cable | 48 in. |
| Length (Metric) | 135 mm |
| Length (Metric) Cable | 1.2 m |
| Material | Glass |
| Show More |