missing translation for 'onlineSavingsMsg'
Learn More

Melford YPD Broth, 1G Capsules

Product Code. 30018960
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 30018960

Brand: Melford Y20091500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Add 50 capsules to 1 liter distilled water and heat until completely dissolved.

Add 50 capsules to 1 liter distilled water and heat until completely dissolved. Sterilize by autoclaving at 121°C for 15 minutes.

Ingredients:

Peptone 20 g/l

Dextrose 20 g/l

Yeast Extract 10 g/l

Specifications

Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.