Learn More
Abcam YOYO™*-1 equivalent
Product Code. 32112525
Change view
Click to view available options
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
32112525 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32112525 Supplier Abcam Supplier No. ab275546200uL
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MW 1270.6 Da. Excellent high affinity counterstain for DNA and chromatin. High fluorescence increase upon binding to nucleic acid. Cell impermeant.
MW 1270.6 Da. Excellent high affinity counterstain for DNA and chromatin. High fluorescence increase upon binding to nucleic acid. Cell impermeant.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 200 μL