missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Yeastolate Solution X50

Product Code. 31908353 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31908353 100 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908353 Supplier Sartorius Supplier No. 013571B
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy

Used to promote the growth of insect cells and enhances the production of recombinant protein.

  • One of the most complete nutritional supplements available
  • Effective in promoting the growth of insect cells
  • Enhances the production of recombinant protein
  • Key component in formulating serum-free media SFM for insect cell culture
  • As peptones, both Yeastolate Solution Concentrate 50X and Lactalbumin hydrolysate LAH are frequently utilized as nutritional supplements for preparing bacterial, insect and mammalian cell culture, especially as a rich source of amino acids.
  • Together in synergy, they both serve as combinations of amino acids, water-soluble peptides, simple and complex carbohydrates CHOs as well as vitamins.
  • Yeastolate is a yeast extract obtained after the autolysis of yeast cells of Saccharomyces cervisiae.
  • Yeastolate is a complex mixture and identification of the constituents responsible for cell growth promotion has yet not been achieved.
  • Further fractionation, isolation and identification of individual active components are needed to better understand the role of these components on cell metabolism.

Specifications

Classification Chemically-defined, Animal Origin-free
Form Liquid
Product Type Cell Culture Supplement
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.