Learn More
Sensititre™ YeastOne™ Broth
Product Code. 15357137
Change view
Click to view available options
Quantity:
11 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15357137 11 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15357137 Supplier Thermo Scientific Oxoid Supplier No. YY3462P
  • ONLINE EXCLUSIVE PRICE
    104.00€
    102.85€ / Pack of 10
    Save 1.15€
    1% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Perform high quality susceptibility testing with supplies including Thermo Scientific™ Sensititre™ YeastOne Broth to enhance your Thermo Scientific™ Sensititre™ test program.
Perform high-quality susceptibility testing with a complete range of supplies, including the Thermo Scientific™ YeastOne Broth, to complement and enhance your Thermo Scientific™ Sensititre™ test program.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Description YeastOne Broth
Product Type Microbiology Media
No. per Pack 10
Packaging Type 11 mL/Bottle
Quantity 11 mL
Species Yeast