Learn More
Melford Yeast with Amino Acids
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
Description
For classifying yeasts based on amino acid and carbohydrates requirements.
For classifying yeasts based on amino acid and carbohydrates requirements. Contains all essential vitamins and inorganic salts necessary for the cultivation of yeasts. Excludes carbohydrate source.
Specifications
Specifications
| Quantity | 500 g |