missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ YARS Recombinant Protein
Brand: Abnova™ H00008565-P02.25ug
Additional Details : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH01933 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
83.82 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MGDAPSPEEKLHLITRNLQEVLGEEKLKEILKERELKIYWGTATTGKPHVAYFVPMSKIADFLKAGCEVTILFADLHAYLDNMKAPWELLELRVSYYENVIKAMLESIGVPLEKLKFIKGTDYQLSKEYTLDVYRLSSVVTQHDSKKAGAEVVKQVEHPLLSGLLYPGLQALDEEYLKVDAQFGGIDQRKIFTFAEKYLPALGYSKRVHLMNPMVPGLTGSKMSSSEEESKIDLLDRKEDVKKKLKKAFCEPGNVENNGVLSFIKHVLFPLKSEFVILRDEKWGGNKTYTAYVDLEKDFAAEVVHPGDLKNSVEVALNKLLDPIREKFNTPALKKLASAAYPDPSKQKPMAKGPAKNSEPEEVIPSRLDIRVGKIITVEKHPDADSLYVEKIDVGEAEPRTVVSGLVQFVPKEELQDRLVVVLCNLKPQKMRGVESQGMLLCASIEGINRQVEPLDPPAGSAPGEHVFVKGYEKGQPDEELKPKKKVFEKLQADFKISEECIAQWKQTNFMTKLGSISCKSLKGGNIS | |
CMTDIC/TYRRS/YRS/YTS | |
YARS | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
8565 | |
YARS (Human) Recombinant Protein (P02) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
YARS | |
Human | |
Recombinant | |
Solution |