Learn More
KARTELL Y Connector
Click to view available options
Diameter (Metric) Inner:
7.6 mm
Unit Size:
Pack of 10
Description
- Autoclavable.
- Dimensions are shown asboth nominal and exact as elasticity of hoses will influence the choice of size.
- Use hoses selector Art.1759 to determine ''grip''. This will then give equivalent crest dimension for the particular connector concerned.
- TESTED ACCORDING TO REACH REGULATION 1907/2006 ANNEX XVII.
- MATERIAL: PP
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 7.6 mm |
| Material | Polypropylene |
| Diameter (English) Outer | 0.47/0.51/0.55 in. |
| Diameter (Metric) Outer | 12, 13, 14 mm |
| Color | White |
| Product Type | Y-Connector |