missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemsplash Xtreme 50 SMS T5/6 Coverall with BlueTaped Seams

Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Description
Chemsplash Xtreme SMS 50 Coverall
- 50GSM Breathable SMS Fabric
- Fully Taped Seams
- Elasticated 3 Pce Hood
- Elasticated Cuffs & Ankles
- Two Way Zip With Adhesive Zip Flap
- Silicone and Latex Free
- PFAS Free
- Comes without the chest label
Description: The Chemsplash Xtreme SMS 50 Coverall with Taped Seams is made from breathable 50gsm SMS fabric, compliant to Cat III Type 5 and 6. It offers a robust and strong barrier to dry hazards and is especially suitable for prolonged periods of use due to its breathable properties. All seams are bound.
Specifications
Specifications
| Product Type | Hooded Coveralls |
| Size | Large |
| Gender | Unisex |
| Material | SMS Fabric |
| Color | Blue |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction