missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ XLKD1 Recombinant Protein

Brand:  Abnova™ H00010894-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195802

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH26231
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
61.16
8
Glutathione Sepharose 4 Fast Flow
10 ug
MARCFSLVLLLTSIWTTRLLVQGSLRAEELSIQVSCRIMGITLVSKKANQQLNFTEAKEACRLLGLSLAGKDQVETALKASFETCSYGWVGDGFVVISRISPNPKCGKNGVGVLIRKVPVSRQFAAYCYNSSDTWTNSCIPEIITTKDPIFNTQTATQTTEFIVSDSTYSVASPYSTIPAPTTTPPAPASTSIPRRKKLICVTEVFMETSTMSTETEPFVENKAAFKNEAAGFGGVPTALLVLALLFFGAAAGLGFCYVKRYVKAFPFTNKNQQKEMIETKVVKEEKANDSNPNEESKKTDKNPEESKSPSKTTVRCLEAEV
CRSBP-1/HAR/LYVE-1/XLKD1
LYVE1
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
10894
XLKD1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LYVE1
Human
Recombinant
Solution
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers