missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ XIAP Tracer mF-Smac

Product Code. 30067458
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 30067458

Brand: Tocris Bioscience™ 8069/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorescein labeled XIAP ligand for TR-FRET assay development

XIAP Tracer mF-Smac is a fluorescent labeled peptide used as an acceptor dye with a terbium labeled BIR domain antibody in TR-FRET assays (Kd values are 4.7 nM at BIR3 (XIAP); 17.9 nM in XIAP polarization assay).
TRUSTED_SUSTAINABILITY

Specifications

Purity 0.95
CAS 783340-88-5
Content And Storage Store at -20°C
Molecular Weight (g/mol) 989.1
Quantity 1 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.