Learn More
Zeiss™ Xenon Flash Bulb
Click to view available options
Type:
Xenon Flash Bulb
Unit Size:
Each
Description
- Ozone free
- Compatible with Axio Imager, Axio Observer, Axio Vert, Axioplan and Axioskop microscopes
Specifications
Specifications
| Type | Xenon Flash Bulb |
| Description | XBO 75W/2 (ozone free) |
| Quantity | 1 Ea. |