missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ XAV 939

Product Code. 30022012
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30022012 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30022012 Supplier R&D Systems™ Supplier No. TB3748GMP/10
  • ONLINE EXCLUSIVE PRICE
    1325.00€
    1249.50€ / 10mg
    Save 75.50€
    5% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. XAV 939 synthesized to cGMP guidelines

XAV 939 synthesized to cGMP guidelines. Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.

Research use only XAV 939 also available.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5,7,8-Tetrahydro-2-[4-(trifluoromethyl)phenyl]-4H-thiopyrano[4,3-d]pyrimidin-4-one
CAS 284028-89-3
Quantity 10 mg
Product Line Tocris
Target Tankyrase Inhibitors
Molecular Formula C14H11F3N2OS
Purity 99%
Regulatory Status GMP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.