Learn More
Thermo Scientific™ Oxoid™ Prepared Brilliant Green Bile Broth D/T Bottled Media
Click to view available options
Fill Weight:
10.0-10.3g
9.5 to 10.5 g
Format:
Poured Tube
Universal bottle
Unit Size:
240mL
Pack of 50
Description
For the detection of coliforms in water and other samples.
Specifications
Specifications
| Color | Mint Green, Transparent |
| Content And Storage | 2°C to 25°C |
| Format | Poured Tube |
| Manufacturing Origin | Germany |
| Product Type | Microbiology Media |
| Fill Weight | 9.5 to 10.5 g |
| Packaging Type | 50 Tubes/Box |
| Quantity | 50/Pk. |