Learn More
Kartell X5 volumetric flask PMP TPXwithcap 1000ml .
Brand:  Kartell 0248900
X5 volumetric flask PMP TPXwithcap 1000ml .
Product Code. 10011111
  • ONLINE EXCLUSIVE PRICE
    172.00€
    146.73€ / Pack of 5
    Save 25.27€
    14% Off
Only null left
Estimated Shipment: 14-04-2026