Learn More
Nalgene Critical Environmental Amber jug with closure 4L
Brand:  Nalgene Critical Environmental 111-04A/C-TOC
Amber jug with closure 4L
Product Code. 15518379
  • ONLINE EXCLUSIVE PRICE
    321.00€
    301.00€ / Each
    Save 20.00€
    6% Off