Learn More
Laboratoires Anios X4 5L SURFANIOS PREMIUM IP STERILE PAE
Brand:  Laboratoires Anios 2409278FJ
X4 5L SURFANIOS PREMIUM IP STERILE PAE
Product Code. 15408464
  • ONLINE EXCLUSIVE PRICE
    338.00€
    306.80€ / Pack of 4
    Save 31.20€
    9% Off
Only null left
Estimated Shipment: 09-06-2026
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • SURFANIOS PREMIUM
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : N-(3-aminopropyl)-N-dodecylpropane-1,3-diamine, didecyldimethylammonium chloride, propan-2-ol