Learn More
Gravis X3 Glass Jar 3L, Clear, Thread 110mm, w/o cap
Brand:  Gravis H0288066LPS
X3 Glass Jar 3L, Clear, Thread 110mm, w/o cap
Product Code. 11869122
  • ONLINE EXCLUSIVE PRICE
    47.70€
    36.84€ / Pack of 3
    Save 10.86€
    22% Off
Only null left
Estimated Shipment: 09-03-2026