Learn More
Ark Plas Products X25 PP Barbed Straight Connector 2.4mm ID
Brand:  Ark Plas Products MAC24-PP0
X25 PP Barbed Straight Connector 2.4mm ID
Product Code. 17856041
  • ONLINE EXCLUSIVE PRICE
    85.89€
    14.71€ / Pack of 25
    Save 71.18€
    82% Off
Only null left
Estimated Shipment: 13-04-2026