Learn More
Kartell X20 graduated beaker low form blue serigraph class B PP 50ml
Brand:  Kartell 0182200
X20 graduated beaker low form blue serigraph class B PP 50ml
Product Code. 12645696
  • ONLINE EXCLUSIVE PRICE
    40.00€
    37.13€ / Pack of 20
    Save 2.87€
    7% Off
Only null left
Estimated Shipment: 28-01-2026