Learn More
Thermo Scientific™ X2 Adapter
Product Code. 12171560
Change view
Click to view available options
Capacity:
50 to 30 mL
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
12171560 50 to 30 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12171560 Supplier Thermo Scientific™ Supplier No. 0101147
Only null left
Add to basket
Add to basket
Adapter 50 mL to 30 mL Oak Ridge
  • X2 Adapter
  • 50 mL Con to 30 mL
TRUSTED_SUSTAINABILITY
Specifications
Capacity 50 to 30 mL
No. per Pack 2/Pk.