Learn More
Fisherbrand X2 350mm Side Arm Shaft, for Wrist Motion Shaker
Brand:  Fisherbrand 88861195
X2 350mm Side Arm Shaft, for Wrist Motion Shaker
Product Code. 15564130
  • ONLINE EXCLUSIVE PRICE
    41.45€
    39.40€ / Pack of 2
    Save 2.05€
    4% Off
Only null left
Estimated Shipment: 16-04-2026