Learn More
Masterflex X12 ISMATEC PUMP TUBING 2-STOP TYGON ® LMT-55, 2.54 mm ID
Brand:  Masterflex 97618-46
133.27 EUR valid until 2026-01-16
BEST PRICE on Promo! Use promo code "24103" to get your promotional price.
X12 ISMATEC PUMP TUBING 2-STOP TYGON ® LMT-55, 2.54 mm ID
Product Code. 16647892
  • 187.00€ / Pack of 12
Outlet Corner
Use promo code 24103
View all promotions
Only null left
Estimated Shipment: 02-03-2026