Learn More
Marienfeld Superior X1000 Precision cover glasses thickness no. 1.5H (
Brand:  Marienfeld Superior 0107222
X1000 Precision cover glasses thickness no. 1.5H (0.17 mm ± 0.005 mm), 24x50 mm, 10x100 pcs./carton
Product Code. 15930396
  • ONLINE EXCLUSIVE PRICE
    477.75€
    236.00€ / Pack of 1000
    Save 241.75€
    50% Off