Learn More
Marienfeld Superior X1000 Precision cover glasses thickness no. 1.5H (
Brand: Marienfeld Superior 0107222
X1000 Precision cover glasses thickness no. 1.5H (0.17 mm ± 0.005 mm), 24x50 mm, 10x100 pcs./carton