Learn More
Marienfeld Superior X1000 Precision cover glasses thickness no. 1.5H (
Brand: Marienfeld Superior 0117500
X1000 Precision cover glasses thickness no. 1.5H (0.17 mm ± 0.005 mm), 10 mm Ø, 10x100 pcs./carton