Learn More
Kimble Chase X1000 Test tube 16x150 WD: 0,6-0,7mm, Boro 5.1glass, without rim, round bottom
Brand:  Kimble Chase A515016000711
150.71 EUR valid until 2025-12-31
BEST PRICE on Promo! Use promo code "24145" to get your promotional price.
X1000 Test tube 16x150 WD: 0,6-0,7mm, Boro 5.1glass, without rim, round bottom
Product Code. 15811628
  • ONLINE EXCLUSIVE PRICE
    225.00€
    150.72€ / Pack of 1000
    Save 74.28€
    33% Off
View all promotions
Only null left
Estimated Shipment: 29-12-2025