Learn More
Kimble Chase X1000 Test tube 13x100 WD: 0,5-0,6mm, Boro 5.1glass, without rim, round bottom
Brand:  Kimble Chase A510013000611
86.59 EUR valid until 2025-12-31
BEST PRICE on Promo! Use promo code "24145" to get your promotional price.
X1000 Test tube 13x100 WD: 0,5-0,6mm, Boro 5.1glass, without rim, round bottom
Product Code. 15881618
  • ONLINE EXCLUSIVE PRICE
    121.00€
    86.60€ / Pack of 1000
    Save 34.40€
    28% Off
View all promotions
Only null left
Estimated Shipment: 30-12-2025