Learn More
Partiguard X100 Overshoes light blue antistatic sole 36 cm
Brand:  Partiguard 55006236
X100 Overshoes light blue antistatic sole 36 cm
Product Code. 30216606
  • ONLINE EXCLUSIVE PRICE
    257.00€
    249.00€ / 100 pairs
    Save 8.00€
    3% Off
Only null left
Estimated Shipment: 16-06-2026