Learn More
Fisherbrand X100 1ml Luer-Slip Plastic Disposable Syringe
Brand:  Fisherbrand 01 36 2448
15.76 EUR valid until 2025-12-31
BEST PRICE on Promo! Use promo code "24159" to get your promotional price.
X100 1ml Luer-Slip Plastic Disposable Syringe
Product Code. 17133105
  • ONLINE EXCLUSIVE PRICE
    17.40€
    16.55€ / Pack of 100
    Save 0.85€
    4% Off