missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Prepared Yeast Extract Agar Bottled Media, 498-502 g, 500 mL sirop bottle, 10 bottles
Click to view available options
Fill Weight:
100.0 to 101.5 g
20 to 20.3 g
498 to 502 g
Format:
125 mL sirop bottle
500 mL Sirop Bottle
Universal Bottle
Unit Size:
1000mL
500mL
Pack of 24
Description
Yeast Extract Agar is used for the enumeration of microorganisms in potable and freshwater samples.
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2°C to 25°C |
| Format | 500 mL Sirop Bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 498 to 502 g |
| Packaging Type | 10 Sirop Bottles with Screw Cap Closures |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction