Learn More
Thermo Scientific™ Violet Red Bile Agar
Click to view available options
Fill Weight:
100 to 101.5 g
Format:
125 mL Sirop Bottle
Unit Size:
Pack of 10
Description
Selectively isolate and enumerate coliform bacteria from food, dairy, or water samples by inhibiting non-coliforms and differentiating lactose fermenters via red colonies using Violet Red Bile Agar.
Specifications
Specifications
| Color | Purple |
| Content And Storage | 2°C to 25°C |
| Format | 125 mL Sirop Bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 100 to 101.5 g |
| Packaging Type | 10 Sirop Bottles with screw cap closures in a box |
| Quantity | 10/Pk. |