Learn More
Thermo Scientific™ Oxoid™ Mueller-Hinton Agar +2% Sodium Chloride
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
Description
Support antimicrobial susceptibility testing with Mueller-Hinton Agar enriched with 2% Sodium Chloride, providing consistent and reliable results.
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2°C to 12°C |
| Format | 90 mm Plate |
| Manufacturing Origin | Germany |
| Product Type | Agar Plate |
| Fill Weight | 17 g |
| Quantity | 10 Plates |