missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 0.1% Peptone Water
Click to view available options
Quantity:
10 x 300 mL
Unit Size:
Pack of 10
Description
Diluent used in the preparation of food and water samples prior to further testing
- Diluent used in the preparation of food and water samples prior to further testing
Specifications
Specifications
| Form | Liquid |
| Manufacturing Origin | United Kingdom |
| Product Type | Broth Tube |
| Sterility | Sterile |
| Target Organism Class | Bacteria |
| Test Type | Microbial Detection |
| Quantity | 10 x 300 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction