missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dichloran-Glycerol (DG18) ISO Agar

Product Code. 18898382 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 18898382

Brand: Thermo Scientific™ PO5388A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Achieve precise fungal isolation with this agar, ideal for targeted microbial growth.

Achieve precise fungal isolation with Dichloran-Glycerol (DG18) ISO Agar, ideal for targeted microbial growth.

TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Content And Storage 2°C to 12°C
Format 90 mm
Manufacturing Origin Germany
Product Type Agar Plate
Fill Volume 17 mL
Quantity 10 Plates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.