Learn More
Corning™ X-SERIES™ Counterweight
Product Code. 17204466 Shop All Corning Products
Click to view available options
Product Type:
Weight: High
Weight: Low
Weight: Medium
Capacity (Metric):
161 to 240 mL
40 to 90 mL
91 to 160 mL
Unit Size:
Each
Product Code. 17204466
Brand: Corning™ 6921
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Corning™ X-SERIES™ Counterweight set is designed for balancing contents of centrifuge prior to centrifugation.
The Corning™ X-SERIES™ Systems are manufactured for Corning™ by ThermoGenesis Corp.
X-SERIES™, X-WASH™, X-LAB™, X-BACS™, and X-MINI™ are either registered trademarks or trademarks of ThermoGenesis Corp. in the United States and/or other countries.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 91 to 160 mL
For Use With (Equipment) X-WASH and X-LAB Systems
Quantity 1/Pk., 1/Cs.
Product Type Weight: Medium