Learn More
Draeger™ X-plore™ 6530 PC Full Face Mask
Product Code. 10310063
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10310063 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10310063 Supplier Draeger™ Supplier No. R55795
  • ONLINE EXCLUSIVE PRICE
    251.00€
    217.51€ / Pack of 1
    Save 33.49€
    13% Off
View all promotions
 Restricted Item
Estimated Shipment: 18-12-2026
Meeting the highest demands for quality and a secure fit, the X-plore™ 6530 mask body is made from EPDM making it highly resistant and skin-friendly, offering high wearing comfort.
  • Safe and comfortable fit
  • Large 180° field of vision
  • Distortion-free lens
  • Efficient ventilation system ensures fog-free lens
  • 5-point harness
  • stainless steel voice diaphragm gives clear voice communication
Specifications
Certifications/Compliance CE, EN 148-1
Certifications (EN) EN 136 Class 3 (CE mark), NIOSH (PC), AS/NZS
Connection Type Standard thread Rd 40 x 1/7 in.
Product Type Full Face Mask
Facepiece Type Personal
Harness Style Adjustable 5-point
Material (Lens) Polycarbonate
Quantity 1 Ea.