Learn More
ABEBA™ X-Light White Lace Up Safety Shoes S2
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Description
- Lightweight, comfort, hygiene
- Comply to EN ISO 20345 : 2012, S2 SRC
- ESD dissipative according to EN 61340 Climate class 1
- Slip resistant PU outsole SRC
- Good cushioning
- White washable microfiber
- Silver ions lining
- Replaceable Active Comfort insole
- Sanitized treatment
Specifications
Specifications
| Certifications/Compliance | CE EN ISO 20345:2012 S2, SRC, ESD |
| Closure Type | Laces |
| Color | White |
| Liner Material | Breathable lining with Silver ions |
| Material | Microfiber |
| Size | 35 |
| Sole Features | Slip and abrasion resistant, shock absorbing, ESD dissipative |
| Sole Material | Polyurethane |
| Style | Lace-up shoes |
| Toe Material | Steel |
| Show More |