Learn More
Thermo Scientific™ X-Gal
Product Code. p-4530703 Shop All Thermo Scientific Products
Click to view available options
Quantity:
0.5 g
1 g
2 x 1 g
Unit Size:
0.5g
1g
2g
Product Code. 10203430
Brand: Thermo Scientific™ R0401
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Effectively screen colonies with X-Gal, an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal, forming an intense blue precipitate.
X-Gal (5-bromo-4-chloro-3-indolyl-beta-D-galacto-pyranoside) is an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal into colorless galactose and 4-chloro-3-brom-indigo, forming an intense blue precipitate. Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies.
Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies. X-Gal Solution is a stable, 0.22μm membrane filtered solution formulated for direct use in conjunction with IPTG for blue/white colony screening.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) formulated for direct use in conjunction with IPTG for blue/white colony screening.
Product Type X-Gal
Quantity 0.5 g