Learn More
Klever X-Change (Wider Cut Head with Scraper)
Product Code. p-7241153
Click to view available options
Color:
Blue
Orange
Red
Yellow
Unit Size:
Pack of 12
Product Code. 17729007
Brand: Klever KCJXC35B
Only null left
Add to basket
Add to basket
Recessed blade of the X-Change (Wider Cut Head with Scraper) reduces cut injuries and damaged goods.
  • Cuts boxes, film, tape and thousands more applications.
  • Ergonomic 'soft touch' handle reduces hand fatigue and stress.
  • Innovative tape splitter.
  • Adavnced plastic polymers.
  • Highest safety level.
  • Interchangeable, replaceable head.
Specifications
For Use With (Application) Cuts Boxes, Film, Tape, and Thousands More Applications
Disposable Reusable
Sterility Non-sterile
Includes Scraper
Color Blue