Learn More
Brady™ Write-On™ Customized Pipemarkers On Card
Click to view available options
Color:
Black
Blue
Brown
Gray
Green
Ochre
Purple
Red
Yellow
Unit Size:
Pack of 10
Description
- Colors conform to BS1710.
Specifications
Specifications
| Certifications/Compliance | DIN VDE 0472 part 815, RoHS compliant using EU Directive 2002/95/EC |
| Description | Write-on pipe markers on card |
| Adhesive Type | Permanent Acrylic, Permanent, Pressure-sensitive |
| Color | Purple |
| Finish | Gloss |
| Format | Card |
| Material | Polyester |
| Product Type | Pipe Marker |
| For Use With (Application) | Pipe and valve marking |
| Labels per Sheet | 10 |
| Show More |