Learn More
Brady™ WorkHorse™ Series Thermal Transfer Printable Labels, B-425
Used to identify property from components to finished products. Brady™ WorkHorse™ Series Thermal Transfer Printable Labels are constructed of permanent polypropylene and is used for thermal transfer printers.
Brand:  Brady™ 031323
 View more versions of this product
Product Code. 11598284
  • ONLINE EXCLUSIVE PRICE
    239.00€
    201.04€ / 1 rolls
    Save 37.96€
    15% Off
Estimated Shipment: 09-12-2026
Description
  • Available in various sizes
Specifications
Specifications
UL Recognized, CSA Accepted, Halogen Free (DIN VDE 0472/part 815)
Acrylic
Matte
Polypropylene
Thermal Transfer Printer Label
Asset Tracking, Barcode Labeling, Component and Equipment Identification, General and Industrial Identification, Rating Plates
1
THT-18-425-3
3000/Pk.
76.20 mm (W) x 25.40 mm (H)
-94°F to 266°F
0.0045 in.
28.58 mm
3 in.
76.20 mm
25.4 mm
Recommended ribbon series: R4300; R4500; R6200; R6400
White
Roll
Thermal Transfer
Non-sterile
BBP81;i5100;i7100;PR Plus;IP Series;THT 76 mm Core Printers;i7500
-70°C
3000 Label(s) / Roll
Rectangle
76.2 x 25.4 mm (W x H)
-70°C to +80°C
0.1143 mm
25.40 x 76.20 mm
76.2 mm
1 in.
THT-18-425-3