Learn More
Wooden Dissecting Board With Frame Edges
Product Code. 13132141
Change view
Click to view available options
Dimensions (L x W x H):
550 x 400 x 17 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
13132141 550 x 400 x 17 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13132141 Supplier S Murray SAMCO Supplier No. Y335/02
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Product Type Dissecting Board
Material Wood
Dimensions (L x W x H) 550 x 400 x 17 mm
Height (Metric) 30 mm