Learn More
Thermo Scientific™ WKITS™ Water QC Kit 1002
Product Code. 12946047
Change view
Click to view available options
Quantity:
3/Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12946047 3/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
If you would like to order, a signed ATCC agreement is required.
Product Code. 12946047 Supplier Thermo Scientific™ Supplier No. R4741002
If you would like to order, a signed ATCC agreement is required.
Thermo Scientific™ WKIT Water QC Kit 1002 is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.
Thermo Scientific™ WKIT Water QC Kit 1002 is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.
  • WaterBugs™ deliver <50 CFUs in a single inoculum to mimic water samples
Ordering Alerts:
Reagents are sufficient for one test/reconstituted vial.
Includes:
Contents: 9 preserved organism vials (3 per organism); 9 rehydration vials. ;Escherichia coli; strain C1, ;Enterobacter cloacae; ATCC™ 13047™,;Proteus mirabilis; ATCC™ 12453™.
Specifications
Description Water QC Kit 1002
Format WKITS
Product Type Quality Control Kit
Quantity 3/Pk.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.