Learn More
Nordvlies WIPEX-STAR wipe for wet cleaning, 80 g/sqm, app. 30x38 cm, z-folded, solvent resistent, blue
Brand:  Nordvlies 1195
WIPEX-STAR wipe for wet cleaning, 80 g/sqm, app. 30x38 cm, z-folded, solvent resistent, blue
Product Code. 15710927
  • ONLINE EXCLUSIVE PRICE
    116.00€
    80.40€ / Pack of 500
    Save 35.60€
    30% Off
Only null left
Estimated Shipment: 13-02-2026